siRNA Details
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026392.1 | NP_080668.1 | 2, 3 | 421 | ||
NM_027415.2 | NP_081691.2 | 2, 3 | 418 | |||
GenBank mRNA | AK003839.1 | - | 2, 3 | 403 | ||
AK005075.1 | - | 2, 3 | 403 | |||
AK008955.1 | - | 2, 3 | 387 | |||
AK151280.1 | - | 2, 3 | 393 | |||
AK167069.1 | - | 2, 3 | 344 | |||
AK169031.1 | - | 2, 3 | 351 | |||
BB860431.1 | - | 2, 3 | 426 | |||
BC011320.1 | - | 2, 3 | 347 | 3979025 |
