siRNA Details
Gene Symbol: Calml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027416.3 | NP_081692.1 | 1 | 279 | ||
GenBank mRNA | AK010118.1 | - | 1 | 277 | ||
BC005457.1 | - | 1 | 274 | 3493620 | ||
EL960451.1 | - | 1 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
