siRNA Details
Gene Symbol: Polr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027423.1 | NP_081699.1 | 6 | 383 | ||
GenBank mRNA | AK039244.1 | - | 6 | 418 | ||
AK039366.1 | - | 6 | 461 | |||
AK165886.1 | - | 6 | 527 | |||
BC068143.1 | - | 6 | 383 | 30362434 |
