siRNA Details
Gene Symbol: Inf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198411.2 | NP_940803.2 | 3 | 533 | ||
GenBank mRNA | CD347830.1 | - | 2 | 166 | ||
DQ834374.1 | - | 3 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
