siRNA Details
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 8 | 862 | ||
GenBank mRNA | AK012014.1 | - | 8 | 853 | ||
AK032897.1 | - | 6 | 401 | |||
AK048488.1 | - | 8 | 885 | |||
AK050391.1 | - | 8 | 862 | |||
AK078856.1 | - | 9 | 821 | |||
AK131608.1 | - | 7 | 773 | |||
AK161812.1 | - | 8 | 837 | |||
AK163839.1 | - | 857 | ||||
AK168006.1 | - | 8 | 864 | |||
AK168752.1 | - | 8 | 884 | |||
AK169128.1 | - | 8 | 863 | |||
BC005755.1 | - | 8 | 817 | 2654151 |
