siRNA Details
Gene Symbol: Ckap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181589.3 | NP_853620.1 | 4 | 981 | ||
GenBank mRNA | AK012040.1 | - | 4 | 933 | ||
AK153766.1 | - | 4 | 940 | |||
BC053443.1 | - | 4 | 933 | 6333921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
