siRNA Details
Gene Symbol: 5730403I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017485.1 | - | 4 | 344 | ||
AK161639.1 | - | 4 | 240 | |||
BC048919.1 | - | 4 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
