siRNA Details
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027439.4 | NP_081715.1 | 9 | 1025 | ||
GenBank mRNA | AB192471.2 | - | 9 | 901 | ||
AK017482.1 | - | 9 | 957 | |||
AK029405.1 | - | 9 | 928 | |||
AK076495.1 | - | 9 | 970 | |||
AK151735.1 | - | 9 | 957 | |||
AK168139.1 | - | 9 | 982 | |||
BC014706.1 | - | 9 | 896 | 4457079 |

More Information
Other Species Targeted

Gene Ontology Categories:
