siRNA Details
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122641.1 | NP_001116113.1 | 18, 19 | 2282 | ||
NM_144529.2 | NP_653112.2 | 19, 20 | 2516 | |||
GenBank mRNA | AB060553.1 | - | 19, 20 | 2514 | ||
AB060554.1 | - | 18, 19 | 2280 | |||
AF073934.1 | - | 1, 2 | 504 | |||
AK147010.1 | - | 19, 20 | 2487 |

More Information
Other Species Targeted

Gene Ontology Categories:
