siRNA Details
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027453.2 | NP_081729.1 | 4, 5 | 458 | ||
GenBank mRNA | AK004239.1 | - | 4, 5 | 489 | ||
AK011367.1 | - | 4, 5 | 804 | |||
AK013120.1 | - | 4, 5 | 446 | |||
AK017614.1 | - | 4, 5 | 419 | |||
AK076283.1 | - | 4, 5 | 458 | |||
BC024612.1 | - | 4, 5 | 441 | |||
BC024920.1 | - | 4, 5 | 401 | |||
BC058282.1 | - | 4, 5 | 414 | 30077136 | ||
BC099407.1 | - | 3, 4 | 393 | 30938785 | ||
BC138215.1 | - | 4, 5 | 439 | 8861234 | ||
BC138216.1 | - | 4, 5 | 439 | 8861235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
