siRNA Details
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081242.2 | NP_001074711.2 | 57 | 7612 | ||
GenBank mRNA | AB093231.1 | - | 31 | 4199 | ||
AK017597.1 | - | 11 | 1429 | |||
AK029828.1 | - | 32 | 4305 | |||
BC033288.1 | - | 19 | 2553 | |||
BC059856.1 | - | 11 | 1373 | |||
BC079896.1 | - | 42 | 5520 | 30544295 | ||
BC158057.1 | - | 57 | 7583 | 9007357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
