siRNA Details
Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027468.1 | NP_081744.1 | 8, 9 | 1112 | ||
GenBank mRNA | AK017670.1 | - | 2, 3 | 342 | ||
BC047389.1 | - | 7, 8 | 1098 | |||
BC055073.1 | - | 8, 9 | 1117 | |||
BC088735.1 | - | 8, 9 | 1117 | |||
BC100404.1 | - | 8, 9 | 1106 | 30471377 |
