siRNA Details
Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027468.1 | NP_081744.1 | 4 | 363 | ||
GenBank mRNA | BC047389.1 | - | 3 | 349 | ||
BC055073.1 | - | 4 | 368 | |||
BC100404.1 | - | 4 | 357 | 30471377 |
