siRNA Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027470.3 | NP_081746.1 | 3 | 413 | ||
GenBank mRNA | AK088512.1 | - | 3 | 390 | ||
AK129298.2 | - | 3 | 409 | |||
AY217016.1 | - | 1 | 128 | |||
BB839524.1 | - | 3 | 387 | |||
BC048238.1 | - | 3 | 375 | 6475114 | ||
CF172418.1 | - | 2 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
