siRNA Details
Gene Symbol: Filip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081243.1 | NP_001074712.1 | 5 | 3862 | ||
GenBank mRNA | BC125581.1 | - | 5 | 3707 | 40130096 | |
BC131965.1 | - | 5 | 3707 | 40130242 |

More Information
Other Species Targeted

Gene Ontology Categories:
