siRNA Details
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080558.4 | NP_542125.3 | 11 | 2287 | ||
GenBank mRNA | AB093303.1 | - | 11 | 2162 | ||
AB120565.1 | - | 11 | 2172 | |||
AK037213.1 | - | 9 | 1997 | |||
AK169543.1 | - | 11 | 2288 | |||
BC008560.1 | - | 4 | 1260 | |||
BC079535.1 | - | 5 | 1666 | |||
BC122519.1 | - | 11 | 2173 | |||
BC141883.1 | - | 11 | 2172 | 40097831 |
