siRNA Details
Gene Symbol: Zdhhc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168516.1 | NP_001161988.1 | 2 | 144 | ||
NM_001168517.1 | NP_001161989.1 | 2 | 191 | |||
NM_027476.3 | NP_081752.2 | 2 | 437 | |||
GenBank mRNA | AK017737.1 | - | 2 | 402 | ||
AK028164.1 | - | 2 | 400 | |||
AK043923.1 | - | 2 | 394 | |||
AK169337.1 | - | 2 | 396 | |||
AK170233.1 | - | 2 | 421 | |||
AK170706.1 | - | 2 | 421 | |||
AK171568.1 | - | 2 | 421 | |||
AK172151.1 | - | 2 | 191 | |||
BC056621.1 | - | 2 | 159 | |||
BC071194.1 | - | 2 | 378 | |||
CA559443.1 | - | 2 | 144 | |||
CD354313.1 | - | 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
