siRNA Details
Gene Symbol: Tmem230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141971.1 | NP_001135443.1 | 4 | 455 | ||
NM_027478.3 | NP_081754.2 | 4 | 515 | |||
GenBank mRNA | AK017729.1 | - | 4 | 510 | ||
AK170062.1 | - | 4 | 488 | |||
BC033429.1 | - | 4 | 486 | 5035633 | ||
BC080755.1 | - | 3 | 1879 | |||
BQ922165.1 | - | 4 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
