siRNA Details
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027490.1 | NP_081766.1 | 6, 7 | 830 | ||
GenBank mRNA | AK017809.1 | - | 6, 7 | 822 | ||
AK152160.1 | - | 6, 7 | 803 | |||
AK153105.1 | - | 6, 7 | 832 | |||
AK153264.1 | - | 6, 7 | 803 | |||
AK153310.1 | - | 6, 7 | 803 | |||
AK166560.1 | - | 6, 7 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
