siRNA Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027495.4 | NP_081771.2 | 2 | 411 | ||
GenBank mRNA | AK017808.1 | - | 2 | 430 | ||
AK079384.1 | - | 2 | 412 | |||
BC018493.1 | - | 2 | 319 | 3593194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
