siRNA Details
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028450.3 | NP_082726.2 | 9, 10 | 986 | ||
GenBank mRNA | AK014093.1 | - | 8, 9 | 735 | ||
AK017798.1 | - | 9, 10 | 964 | |||
AK028490.1 | - | 8, 9 | 902 | |||
AK036670.1 | - | 9, 10 | 987 | |||
BC032154.1 | - | 9, 10 | 1157 | 5068339 | ||
BY713087.1 | - | 8, 9 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
