siRNA Details
Gene Symbol: Fam175a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172405.3 | NP_765993.1 | 9 | 1178 | ||
GenBank mRNA | AK014420.1 | - | 10 | 1268 | ||
AK042001.1 | - | 10 | 1252 | |||
AK042339.1 | - | 10 | 1252 | |||
AK051816.1 | - | 9 | 1179 | |||
AK167540.1 | - | 9 | 1209 | |||
BC029845.1 | - | 9 | 1183 | 4010744 | ||
BC043923.1 | - | 8 | 1097 |
