siRNA Details
Gene Symbol: Fam175a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172405.3 | NP_765993.1 | 7 | 649 | ||
GenBank mRNA | AK042001.1 | - | 7 | 658 | ||
AK042339.1 | - | 7 | 658 | |||
AK051816.1 | - | 7 | 650 | |||
AK167540.1 | - | 7 | 680 | |||
BC029845.1 | - | 7 | 654 | 4010744 | ||
BC043923.1 | - | 6 | 563 |
