siRNA Details
Gene Symbol: Utp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175158.3 | NP_780367.2 | 52 | 7008 | ||
GenBank mRNA | AB052760.1 | - | 10 | 1210 | ||
AK047081.1 | - | 1059 | ||||
BC005522.1 | - | 10 | 1230 | |||
BC048955.1 | - | 22 | 2873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
