siRNA Details
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081205.1 | NP_001074674.1 | 5 | 594 | ||
GenBank mRNA | AK014427.1 | - | 5 | 547 | ||
AK075654.1 | - | 5 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
