siRNA Details
Gene Symbol: Angptl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145154.2 | NP_660136.1 | 5 | 1279 | ||
GenBank mRNA | AB054065.1 | - | 5 | 1279 | ||
AK018113.1 | - | 4 | 648 | |||
BC025904.1 | - | 5 | 1257 | 5049765 |

More Information
Other Species Targeted

Gene Ontology Categories:
