siRNA Details
Gene Symbol: Rasl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033158.2 | NP_001028330.2 | 5 | 1799 | ||
GenBank mRNA | AK014511.1 | - | 5 | 1762 | ||
AK171623.1 | - | 5 | 1755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
