siRNA Details
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153551.1 | NP_705779.1 | 24 | 2264 | ||
GenBank mRNA | AK040537.1 | - | 21 | 2578 | ||
BC035208.1 | - | 24 | 2264 | 3671384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
