siRNA Details
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081359.3 | NP_001074828.2 | 54 | 8043 | ||
NM_001112721.2 | NP_001106192.1 | 54 | 8025 | |||
GenBank mRNA | AK014485.1 | - | 3 | 1205 | ||
AK122398.1 | - | 31 | 4516 | |||
AY550908.1 | - | 54 | 7633 | |||
BC034075.1 | - | 3 | 252 | |||
BC047348.1 | - | 45 | 6536 | |||
BC049162.1 | - | 25 | 3723 | |||
BC057458.1 | - | 7 | 1062 | |||
BC057923.1 | - | 6 | 575 | |||
BC064751.1 | - | 4 | 285 |

More Information
Other Species Targeted

Gene Ontology Categories:
