siRNA Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175160.3 | NP_780369.1 | 6 | 819 | ||
GenBank mRNA | AK014489.1 | - | 1 | 1687 | ||
AK038593.1 | - | 6 | 821 | |||
BC026570.1 | - | 2 | 400 | 4984252 | ||
BC119378.1 | - | 3 | 558 | 8734127 | ||
BC144878.1 | - | 3 | 558 | 9053426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
