siRNA Details
Gene Symbol: Pwwp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164231.1 | NP_001157703.1 | 2 | 1285 | ||
NM_027557.1 | NP_081833.1 | 2 | 1285 | |||
GenBank mRNA | AK014534.1 | - | 1 | 245 | ||
AK171257.1 | - | 3 | 1478 | |||
AK173305.1 | - | 2 | 1277 |

More Information
Other Species Targeted

Gene Ontology Categories:
