siRNA Details
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027558.1 | NP_081834.1 | 2 | 522 | ||
GenBank mRNA | AK014543.1 | - | 2 | 421 | ||
AK140064.1 | - | 2 | 502 | |||
AX775753.1 | - | 2 | 478 | |||
BC044759.2 | - | 2 | 463 | |||
CD363842.1 | - | 2 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
