siRNA Details
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 2 | 105 | ||
NM_001168224.1 | NP_001161696.1 | 2 | 217 | |||
NM_027562.4 | NP_081838.1 | 3 | 279 | |||
GenBank mRNA | AK014570.1 | - | 3 | 280 | ||
AK137753.1 | - | 2 | 215 | |||
AY137341.1 | - | 2 | 217 | |||
AY137342.1 | - | 2 | 105 | |||
BC117543.1 | - | 3 | 269 | 40105656 | ||
JF792238.1 | - | 2 | 116 | |||
JF792239.1 | - | 2 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
