siRNA Details
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 2, 3 | 114 | ||
NM_001168224.1 | NP_001161696.1 | 2, 3 | 226 | |||
NM_027562.4 | NP_081838.1 | 3, 4 | 288 | |||
GenBank mRNA | AK014570.1 | - | 3, 4 | 289 | ||
AK137753.1 | - | 2, 3 | 224 | |||
AY137341.1 | - | 2, 3 | 226 | |||
AY137342.1 | - | 2, 3 | 114 | |||
BC117543.1 | - | 3, 4 | 278 | 40105656 | ||
JF792238.1 | - | 2, 3 | 125 | |||
JF792239.1 | - | 2, 3 | 176 |
