siRNA Details
Gene Symbol: Ccdc93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025156.2 | NP_001020327.1 | 3 | 351 | ||
NM_029955.3 | NP_084231.2 | 3 | 351 | |||
GenBank mRNA | AK014621.1 | - | 3 | 325 | ||
AK020306.1 | - | 3 | 311 | |||
AK052300.1 | - | 3 | 316 | |||
AK082581.1 | - | 3 | 344 | |||
AK085701.1 | - | 3 | 310 | |||
AK142302.1 | - | 3 | 369 | |||
AK159415.1 | - | 3 | 319 | |||
BC054083.1 | - | 3 | 254 | 6467830 | ||
BY136404.1 | - | 3 | 351 | |||
BY730410.1 | - | 3 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
