siRNA Details
Gene Symbol: Lrrc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027581.1 | NP_081857.1 | 3 | 841 | ||
GenBank mRNA | AK014850.1 | - | 2 | 573 | ||
BC145796.1 | - | 3 | 839 | 40131099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
