siRNA Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027588.3 | NP_081864.2 | 3 | 275 | ||
GenBank mRNA | AF356186.1 | - | 3 | 248 | ||
AJ295253.1 | - | 3 | 202 | |||
AK014892.1 | - | 3 | 275 | |||
BC060976.1 | - | 3 | 267 | 6773248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
