siRNA Details
Gene Symbol: Ints10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293791.1 | NP_001280720.1 | 1, 2 | 221 | ||
NM_001293792.1 | NP_001280721.1 | 1, 2 | 221 | |||
NM_027590.4 | NP_081866.1 | 1, 2 | 221 | |||
GenBank mRNA | AK014939.1 | - | 1, 2 | 197 | ||
AK030974.1 | - | 1, 2 | 184 | |||
AK034498.1 | - | 1, 2 | 137 | |||
AK040574.1 | - | 1, 2 | 222 | |||
AK047093.1 | - | 1, 2 | 216 | |||
AK083331.1 | - | 1, 2 | 176 | |||
AK147743.1 | - | 1, 2 | 195 | |||
AK169819.1 | - | 1, 2 | 196 | |||
BC031923.1 | - | 1, 2 | 184 | 4486323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
