siRNA Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014905.1 | - | 10 | 1437 | ||
AM690750.1 | - | 9 | 1078 | |||
AM690751.1 | - | 9 | 1078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
