siRNA Details
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081046.1 | NP_001074515.1 | 4 | 558 | ||
GenBank mRNA | AK014861.1 | - | 4 | 552 | ||
AK162207.1 | - | 13 | 1043 | |||
BC050801.1 | - | 4 | 556 | 6770929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
