siRNA Details
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081046.1 | NP_001074515.1 | 6 | 772 | ||
GenBank mRNA | BC050801.1 | - | 6 | 770 | 6770929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
