siRNA Details
Gene Symbol: 4921524L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027598.1 | NP_081874.1 | 6, 7 | 666 | ||
GenBank mRNA | AK014961.1 | - | 6, 7 | 667 | ||
BC049627.1 | - | 4, 5 | 511 | 6706381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
