siRNA Details
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271397.1 | NP_001258326.1 | 11 | 3331 | ||
GenBank mRNA | AK017551.1 | - | 4 | 407 | ||
AK029961.1 | - | 11 | 3044 | |||
AK147172.1 | - | 11 | 3067 | |||
BC049166.1 | - | 11 | 2921 | |||
BC058699.1 | - | 11 | 3487 | |||
BC094941.1 | - | 3 | 201 | |||
BC125575.1 | - | 11 | 3057 | 40130090 | ||
BC125579.1 | - | 11 | 3057 | 40130094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
