siRNA Details
Gene Symbol: 4921539E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163494.1 | NP_001156966.1 | 3 | 797 | ||
NM_027612.1 | NP_081888.1 | 4 | 889 | |||
GenBank mRNA | AK015018.1 | - | 4 | 884 | ||
AK161421.1 | - | 4 | 889 | |||
BC107404.1 | - | 3 | 738 | 40058776 | ||
BC107405.1 | - | 4 | 830 | 40058778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
