siRNA Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163507.1 | NP_001156979.1 | 8 | 3539 | ||
NM_027613.1 | NP_081889.1 | 8 | 3542 | |||
GenBank mRNA | AJ586616.1 | - | 3 | 512 | ||
AK036724.1 | - | 8 | 3381 | |||
AK137556.1 | - | 8 | 3379 | |||
BC046425.1 | - | 3 | 842 | |||
BC137623.1 | - | 8 | 3342 | 8860642 | ||
BC144789.1 | - | 8 | 3339 | 9053337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
