siRNA Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163507.1 | NP_001156979.1 | 6 | 1371 | ||
NM_027613.1 | NP_081889.1 | 6 | 1371 | |||
GenBank mRNA | AK036724.1 | - | 6 | 1209 | ||
AK137556.1 | - | 6 | 1210 | |||
BC137623.1 | - | 6 | 1171 | 8860642 | ||
BC144789.1 | - | 6 | 1171 | 9053337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
