siRNA Details
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027617.3 | NP_081893.1 | 12 | 1401 | ||
GenBank mRNA | BC139279.1 | - | 11 | 1314 | 8862301 | |
BC139280.1 | - | 11 | 1314 | 8862302 | ||
BC171970.1 | - | 11 | 1312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
