siRNA Details
Gene Symbol: Poteg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026256.2 | NP_080532.1 | 1 | 263 | ||
NM_027618.1 | NP_081894.1 | 1 | 263 | |||
GenBank mRNA | AK015012.1 | - | 1 | 264 | ||
AK015165.1 | - | 1 | 264 | |||
AK015473.1 | - | 1 | 264 | |||
DQ298444.1 | - | 1 | 264 | |||
DQ517440.1 | - | 1 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
