siRNA Details
Gene Symbol: Eva1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199210.1 | NP_001186139.1 | 6, 7 | 1265 | ||
NM_027627.2 | NP_081903.1 | 5, 6 | 1121 | |||
GenBank mRNA | AF358257.1 | - | 6, 7 | 992 | ||
AK016443.1 | - | 8, 9 | 1261 | |||
AK037486.1 | - | 4, 5 | 976 | |||
BC100350.1 | - | 5, 6 | 867 | 30522196 | ||
BC132305.1 | - | 6, 7 | 1265 | 40130582 | ||
BC144751.1 | - | 5, 6 | 1067 | |||
BC157969.1 | - | 4, 5 | 815 | 9007291 |
