siRNA Details
Gene Symbol: Atoh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153778.3 | NP_722473.1 | 1 | 1011 | ||
GenBank mRNA | AB046527.1 | - | 1 | 1013 | ||
AB046528.1 | - | 1 | 709 | |||
AB049066.1 | - | 1 | 598 | |||
AY349615.1 | - | 1 | 719 | |||
BC023684.1 | - | 1 | 679 | 5343472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
